missing translation for 'onlineSavingsMsg'
Learn More

Medisca Atenolol USP

Catalog No. 14186453
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186453 25 g
14185286 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14186453 Supplier Medisca Supplier No. 026204*
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Atenolol USP is a white to practically white, odorless powder.

Atenolol USP is a white to practically white, odorless powder.

Specifications

Chemical Name or Material Atenolol USP
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 29122-68-7
Type Drug Standard
Molecular Weight (g/mol) 266.3388
Quantity 25 g
Percent Purity 98 to 102 %
Grade USP