missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Atenolol

Catalog No. AAJ6119903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6119903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6119903 Supplier Thermo Scientific Chemicals Supplier No. J6119903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29122-68-7 | C14H22N2O3 | 266.341 g/mol

Cardioselective beta-adrenergic blocker. An antihypertensive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Cardioselective beta-adrenergic blocker
  • An antihypertensive

Chemical Identifiers

CAS 29122-68-7
Molecular Formula C14H22N2O3
Molecular Weight (g/mol) 266.341
MDL Number MFCD00057645
InChI Key METKIMKYRPQLGS-UHFFFAOYSA-N
Synonym atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
PubChem CID 2249
ChEBI CHEBI:2904
IUPAC Name 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
SMILES CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O

Specifications

Melting Point 151°C to 155°C
Color White
Quantity 1 g
Merck Index 14,859
Formula Weight 266.34
Physical Form Powder
Chemical Name or Material (+/-)-Atenolol
EINECSNumber 249-451-7
RTECSNumber AC3600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only