missing translation for 'onlineSavingsMsg'
Learn More

Atazanavir for system suitability, MilliporeSigma™ Supelco™

Catalog No. 111002260
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002260 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002260 Supplier Merck Emd Millipore Supplier No. Y0002064

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Specifications

CAS 198904-31-3
Linear Formula C38H52N6O7
MDL Number MFCD08435966
Synonym Atazanavir; 1,14-Dimethyl (3 S,8 S,9 S,12 S)-3,12-bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-pentaazatetradecanedioate; BMS-232632
Type Pharmacopeia and Metrological Institutes Standards
Quantity 10 mg
Formula Weight 704.86
Grade Pharmaceutical Primary Standard