missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D5442 C12-C60 Quantitative Linearity Standard, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022610
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022610 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022610 Supplier Merck Emd Millipore Supplier No. 502243

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ASTM(R) D5442 Quantitative Linearity Standard consists of mixes of n-paraffins used for determining column resolution, retention times, response factors, and internal standard preparation. Each component is 0.01% (w/w) in cyclohexane.

Specifications

Flash Point -18°C (Closed Cup)
UN Number UN1145 - Class 3 - PG 2 - Cyclohexane
Quantity Each
Grade Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials