missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D5441 MTBE Contaminants (High) Mix A, Certified Reference Material, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022559
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022559 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022559 Supplier Merck Emd Millipore Supplier No. 47942

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
Flash Point -33°C (Closed Cup)
UN Number UN 2398 3 / PGII
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials