missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ D5307 Crude Oil, Quantitative Standard 6.25% (w/w) Each Component, MilliporeSigma™ Supelco™

Catalog No. 111004629
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004629 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004629 Supplier Merck Emd Millipore Supplier No. SGMA48179

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Equal weights of the hydrocarbons listed. Use to determine detector response factors (slight warming of mixture may be necessary to dissolve all components completely.)

Specifications

Packaging Ampule
Concentration 6.25% (w/w) each component
Quantity 2 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Decane; Dodecane; Dotriacontane; Eicosane; Heptadecane; Hexadecane; Hexatriacontane; Octacosane; Octadecane; Pentadecane; Tetracontane; Tetracosane; Tetradecane; Tetratetracontane; Tridecane; Undecane

ASTM is a registered trademark of American Society for Testing and Materials