missing translation for 'onlineSavingsMsg'
Learn More

ASTM™ 2-Hexanone, MilliporeSigma™ Supelco™

Catalog No. 111017913
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017913 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017913 Supplier Merck Emd Millipore Supplier No. 47733U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Hexanone is a colorless to pale yellow liquid with sharp odor belonging to the class of ketones. It is obtained as a waste product of coal gasification, oil shale operations and wood pulping. It is widely used as a solvent in industry.

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Petroleum

Specifications

Product Line ASTM
CAS 591-78-6
Vapor Pressure 10 mmHg (39°C)
Density 0.812 g/mL (at 25°C)
Flash Point 23°C
Physical Form Neat
Linear Formula CH3(CH2)3 COCH3
MDL Number MFCD00009482
Melting Point -57°C
Boiling Point 127°C
Synonym Butyl methyl ketone
Type Food and Cosmetic Component Standard
Quantity 5 mL
Formula Weight 100.16
Grade Analytical Standard
Show More Show Less

ASTM is a registered trademark of American Society for Testing and Materials