missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Astec™ CYCLOBOND™ I 2000 RSP Chiral HPLC Guard Cartridge, 5 μm Particle Size

Catalog No. 111009699
Change view
Click to view available options
Diameter: 4 mm; Length: 2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111009699 Diameter: 4 mm; Length: 2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009699 Supplier MilliporeSigma™ Supelco™ Supplier No. 21103AST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Astec™ CYCLOBOND™ I 2000 RSP chiral HPLC column is basically comprised of native and derivatized β and γ cyclodextrins. It has been used for numerous chiral separations via selectively trapping the organic molecules in the cyclodextrin cavity.

The hydroxyl groups on the surface of the beta-cyclodextrin are reacted with (R,S)-propylene oxide to yield a general purpose chiral stationary phase. It has the added property of separating non-aromatic structures such as t-boc amino acids, for which it is a standard methodology.

Specifications

Quantity 1/Pk.
Particle Size 5 μm
Length (Metric) 2 cm
Type Chiral HPLC Guard Cartridge
Diameter (Metric) Inner 4 mm
Phase Chiral
For Use With (Application) HPLC
Product Line Astec CYCLOBOND

Astec and CYCLOBOND are registered trademarks of Sigma-Aldrich Co. LLC