missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Astec™ CHIRALDEX™ B-DM Capillary GC Column, df 0.12 μm

Used for enantiomeric separation of ephedrine, pseudoephedrine, chlorinated intermediates and methylamphetamine by gas chromatographic analysis.

Supplier:  MilliporeSigma™ Supelco™ 77025AST

 View more versions of this product

Catalog No. 111005894


Only null left
Explore available promotions

Description

Description

Through special derivatization techniques, the concentration of the cyclodextrin in the CHIRALDEX™ B-DM has been substantially increased in the polysiloxane carrier. This phase is very useful for a number of free acids and bases. The B-DM is able to perform most of the separations done on a beta-permethylated phase, but with higher resolution. The selectivity of the B-DM covers applications of both the B-PM and B-PH phases, although with superior performance.
Specifications

Specifications

0.25 mm
Astec CHIRALDEX
0.12 μm
Fused Silica
Capillary GC
Gas Chromatography (GC)
50 m
2,3-di-O-methyl-6-t-butyl silyl derivative of β-cyclodextrin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Astec and CHIRALDEX are registered trademarks of Sigma-Aldrich Co. LLC