missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips

AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips. Ideal for procedures that require a high level of precision and tactile sensitivity. DermaSense technology helps reduce the risk of skin reactions. Textured fingertips ensure a secure grip. Chemo-tested.

$182.27 - $182.27

Specifications

Chlorinated No
Accelerator Free Yes
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17101078
View Documents
Medicom™
9970A
X-Small
Case of 2000 Each for $182.27
Only null left
 
17101079
View Documents
Medicom™
9970B
Small
Case of 2000 Each for $182.27
Only null left
 
17101080
View Documents
Medicom™
9970C
Medium
Case of 2000 Each for $182.27
Only null left
 
17101081
View Documents
Medicom™
9970D
Large
Case of 2000 Each for $182.27
Only null left
 
17101082
View Documents
Medicom™
9970E
X-Large
Case of 2000 Each for $182.27
Only null left
 
Description

Description

  • These chemo-tested, ultra-thin nitrile gloves are ideal for procedures that require precision and tactile sensitivity.
  • Textured fingertips ensure a secure grip, even when wet.
  • DermaSense technology eliminates the chemical accelerators commonly found in nitrile gloves to reduce the risk of skin reactions.
Specifications

Specifications

No
Healthcare, Industrial, Laboratory
Blue
Fingertip Textured
Powder Free
Yes
Yes
1.5
Single-use
Yes
Non-sterile
Beaded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'