missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 17101080
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17101080 Medium
17101079 Small
17101081 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17101080 Supplier Medicom™ Supplier No. 9970C
Edge

AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips. Ideal for procedures that require a high level of precision and tactile sensitivity. DermaSense technology helps reduce the risk of skin reactions. Textured fingertips ensure a secure grip. Chemo-tested.

  • These chemo-tested, ultra-thin nitrile gloves are ideal for procedures that require precision and tactile sensitivity.
  • Textured fingertips ensure a secure grip, even when wet.
  • DermaSense technology eliminates the chemical accelerators commonly found in nitrile gloves to reduce the risk of skin reactions.

Specifications

Certifications/Compliance Chemo Tested, ISO 13485:2016
Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free Yes
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (English) 2.76 mil
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model 9970-C
Quantity 200/Box
Show More Show Less