missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Power™ Nitrile Medical Examination Gloves with Textured Fingertips

AssureTouch™ Power™ Nitrile Medical Examination Gloves with Textured Fingertips. Provides strong puncture and tear resistance without compromising tactility. Extended cuffs provide optimal coverage. Textured fingertips for a secure grip, even when wet. Chemo tested and fentanyl safe.

$17.34 - $17.34

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17101073
View Documents
Medicom™
9999A
X-Small
Pack of 100 Each for $17.34
Case of 10 Pack for $173.40
Only null left
 
17101074
View Documents
Medicom™
9999B
Small
Pack of 100 Each for $17.34
Case of 10 Pack for $173.40
Only null left
 
17101075
View Documents
Medicom™
9999C
Medium
Pack of 100 Each for $17.34
Case of 10 Pack for $173.40
Only null left
 
17101076
View Documents
Medicom™
9999D
Large
Pack of 100 Each for $17.34
Case of 10 Pack for $173.40
Only null left
 
17101077
View Documents
Medicom™
9999E
X-Large
Pack of 100 Each for $17.34
Case of 10 Pack for $173.40
Only null left
 
Description

Description

  • These gloves provide strong puncture, tear and chemical resistance without compromising dexterity or tactility.
  • Extended cuffs provide optimal coverage.
  • Textured fingertips ensure a secure grip, even when wet.
  • Chemo tested and fentanyl safe.
Specifications

Specifications

No
Healthcare, Industrial, Laboratory
Blue
Fingertip Textured
Powder Free
Yes
No
1.5
Single-use
Yes
Non-sterile
Beaded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'