missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Power™ Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 17101077
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17101077 X-Large
17101073 X-Small
17101074 Small
17101075 Medium
17101076 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17101077 Supplier Medicom™ Supplier No. 9999E
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AssureTouch™ Power™ Nitrile Medical Examination Gloves with Textured Fingertips. Provides strong puncture and tear resistance without compromising tactility. Extended cuffs provide optimal coverage. Textured fingertips for a secure grip, even when wet. Chemo tested and fentanyl safe.

  • These gloves provide strong puncture, tear and chemical resistance without compromising dexterity or tactility.
  • Extended cuffs provide optimal coverage.
  • Textured fingertips ensure a secure grip, even when wet.
  • Chemo tested and fentanyl safe.

Specifications

Certifications/Compliance FDA 510k, Chemo Tested, Fentanyl safe
Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Sterility Non-sterile
Thickness (English) 4 mil
Thickness (Metric) 0.1016 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Quantity 100/Box
Show More Show Less