missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Basics Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips

AssureTouch™ Basics Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips. These affordable gloves provide healthcare professionals with hand protection for handling chemotherapy drugs. High-quality nitrile won’t compromise tactile sensitivity and ensure a secure grip.

$7.79 - $7.79

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17101088
View Documents
Medicom™
9985A
X-Small
Pack of 100 Each for $7.79
Case of 10 Pack for $77.90
Only null left
 
17101089
View Documents
Medicom™
9985B
Small
Pack of 100 Each for $7.79
Case of 10 Pack for $77.90
Only null left
 
17101090
missing translation for 'viewDocuments'
Medicom™
9985C
Medium
Pack of 100 Each for $7.79
Case of 10 Pack for $77.90
Only null left
 
17101091
missing translation for 'viewDocuments'
Medicom™
9985D
Large
Pack of 100 Each for $7.79
Case of 10 Pack for $77.90
Only null left
 
17101092
missing translation for 'viewDocuments'
Medicom™
9985E
X-Large
Pack of 100 Each for $7.79
Only null left
 
Description

Description

  • These affordable chemo-tested gloves provide healthcare professionals with the hand protection they need to prevent exposure and contamination when handling chemotherapy drugs.
  • High-quality nitrile won’t compromise manual dexterity or tactile sensitivity and textured fingertips ensure a secure grip, even when wet.
Specifications

Specifications

No
Healthcare, Industrial, Laboratory
Blue
Fingertip Textured
Powder Free
Yes
No
1.5
Single-use
Yes
Non-sterile
Beaded
Videos
SDS
Documents

Documents