missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Basics Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips

Catalog No. 17101088
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17101088 X-Small
17101089 Small
17101090 Medium
17101091 Large
17101092 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17101088 Supplier Medicom™ Supplier No. 9985A
Edge

AssureTouch™ Basics Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips. These affordable gloves provide healthcare professionals with hand protection for handling chemotherapy drugs. High-quality nitrile won’t compromise tactile sensitivity and ensure a secure grip.

  • These affordable chemo-tested gloves provide healthcare professionals with the hand protection they need to prevent exposure and contamination when handling chemotherapy drugs.
  • High-quality nitrile won’t compromise manual dexterity or tactile sensitivity and textured fingertips ensure a secure grip, even when wet.

Specifications

Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 2.8 mil
Latex Free Yes
Material Nitrile
Palm Thickness (English) 1.97 mil
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model 9985-A
Quantity 1000/Cs.
Show More Show Less