missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Basics Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips

Catalog No. 17101089
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17101089 Small
17101088 X-Small
17101090 Medium
17101091 Large
17101092 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17101089 Supplier Medicom™ Supplier No. 9985B
Only null left

AssureTouch™ Basics Nitrile Powder-Free Medical Examination Gloves with Textured Fingertips. These affordable gloves provide healthcare professionals with hand protection for handling chemotherapy drugs. High-quality nitrile won’t compromise tactile sensitivity and ensure a secure grip.

  • These affordable chemo-tested gloves provide healthcare professionals with the hand protection they need to prevent exposure and contamination when handling chemotherapy drugs.
  • High-quality nitrile won’t compromise manual dexterity or tactile sensitivity and textured fingertips ensure a secure grip, even when wet.

Specifications

Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 2.8 mil
Latex Free Yes
Material Nitrile
Palm Thickness (English) 1.97 mil
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model 9985-B
Quantity 1000/Cs.
Show More Show Less