missing translation for 'onlineSavingsMsg'
Learn More

Associated Bag Co. HazMat Bags

Catalog No. 19821939
Encompass
Change view
Click to view available options
Material:
LDPE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19821939 LDPE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19821939 Supplier Associated Bag Supplier No. 308100Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bold colors and printed warnings identify hazardous or contaminated contents

  • Several message options available
  • Strong bottom seal
  • 38 × 60 in. (96.5 x 152.4cm) hazardous materials low-density flat liner
  • Free of heavy metals and safe for landfill or incineration according to CONEG (Coalition of Northeastern Governors) legislation regarding the use of heavy metals in packaging

Specifications

Color Yellow
Includes Biohazard label
For Use With (Application) Safe for landfill or incineration
Thickness 6 mil
Material LDPE
Product Type Hazardous Materials Low Density Flat Liner