missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials

Catalog No. 0337518D
Change view
Click to view available options
Color:
Black
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0337518D Natural
03378352 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0337518D Supplier Thermo Scientific™ Supplier No. C40135A

Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.

Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.

  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles

Specifications

Description PP, Natural; Integral PP septum
Color Natural
Material Polypropylene
Septum Integral Polypropylene
Closure Size 8-425
For Use With (Equipment) 8-425 Standard Opening Vials
Product Type SepCap