missing translation for 'onlineSavingsMsg'
Learn More

AsCpf1, Mouse anti-Bacteria, Clone: 3D3-F7, Millipore Sigma™

Numéro de catalogue. p-7162906
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MABE182325 25 μg
MABE1823100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MABE182325 Fournisseur MilliporeSigma Code fournisseur MABE182325UG
Il en reste null

Mouse Monoclonal Antibody

Anti-AsCpf1, clone 3D3-F7, Cat. No. MABE1823, is a highly specific mouse monoclonal antibody that targets CRISPR-associated endonuclease Cpf1 and has been tested in Western Blotting.

CRISPR-associated endonuclease Cas12a (UniProt: U2UMQ6; also known as EC: 3.1.27.2, AsCpf1, CRISPR-associated endonuclease Cpf1) I encoded by the Cas12a (also known as cpf1) gene in Acidaminococcus species. CRISPR is an adaptive immune system that provides protection against mobile genetic elements, such as viruses, transposable elements, and conjugative plasmids. CRISPR clusters contain sequences complementary to antecedent mobile elements and target invading nucleic acids. They are transcribed and processed into CRISPR RNA (crRNA). Cas12a, an endonuclease from the type V-A CRISPR system, has several unique characteristics when compared to Cas9 system that increase the utility of CRISPR-based gene editing. Cas12a mediated cleavage utilizes a single and short CRISPR RNA) without requiring trans-activating crRNA. Cas12a recognize a 5' T-rich protospacer adjacent motif (PAM, TTTN for this specific enzyme), unlike Cas9 enzymes which recognize 3' G-rich PAMs, thus this enzyme increases the possibilities for genome editing. The endonuclease activity of Cas12a results in staggered 4-base 5' overhangs 19 and 22 bases downstream of the PAM on the non-targeted and targeted strand respectively.

Spécifications

Antigen AsCpf1
Applications Western Blot
Classification Monoclonal
Clone 3D3-F7
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected AsCpf1 in HEK293 cells expressing HA-tagged AsCpf1 (Courtesy of Stefan Schuchner, Ph.D. and Egon Ogris, M.D., Medical University of Vienna, Austria).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias EC 3.1.27.2;AsCpf1;CRISPR-associated endonuclease Cpf1
Gene Symbols Cas12a;Cpf1
Host Species Mouse
Immunogen His-tagged recombinant fragment corresponding to 197 amino acids from the C-terminal region of Acidaminococcus Cas12a.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 3D3-F7 specifically detects CRISPR-associated endonuclease Cas12a.
Target Species Bacteria
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats