missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbyl Palmitate, NF, 95-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Ascorbyl Palmitate, NF is often used to promote flow of tableting powders and to prevent their sticking in tablet presses. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.1% |
| Loss on Drying | 2% |
| Packaging | Poly Bottle |
| Quantity | 50 kg |
| Specific Rotation | +21° to +24° |
| Percent Purity | 95 to 100.5% |
| Grade | NF |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Ascorbyl Palmitate |