Learn More
MilliporeSigma Ascentis™ Express PCS-Phenyl-Hexyl HPLC Column 3 Pack
Ascentis™ Express PCS-Phenyl-Hexyl columns are specifically engineered to provide efficient separation of basic, acidic, or neutral compounds under low ionic strength (formic acid) mobile phase conditions.
Supplier:  MilliporeSigma 50704U
 View more versions of this product
Catalog No. 01-000-894
Only null left
Explore available promotions
Description
Description
These columns contain advanced Fused-Core™ particles with pores strategically sized to 90 Å for small molecule analyses. This attribute makes these columns an excellent choice for pharmaceutical method development.
  • The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic compounds as compared to traditional C18 chemistries.
  • This column also provides orthogonal selectivity to other small molecule chemistries.
Specifications
Specifications
3.0 mm
2.7 μm
Positively Charged Surface (PCS)
HPLC Column
5 mm
90 Å
Stainless Steel
HPLC
Product Suggestions
Product Suggestions