missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASB3 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. 01675545
Modifier l'affichage
Click to view available options
Quantité:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
01675545 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01675545 Fournisseur Invitrogen™ Code fournisseur PA141416
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

This antibody will not cross-react with mouse or rat asb-3. Suggested positive control: human spleen protein, antigen standard for ASB3 (transient overexpression lysate), human spleen.

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known.

Spécifications

Antigène ASB3
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugué Unconjugated
Formule PBS with 100μg/mL BSA and 0.02% sodium azide
Gène ASB3
Numéro d’acquisition du gène Q9WV72, Q9Y575
Alias du gène 2400011J03Rik; ankyrin repeat and SOCS box containing 3; ankyrin repeat and SOCS box protein 3; ankyrin repeat and SOCS box-containing 3; ankyrin repeat and SOCS box-containing 3-like; ankyrin repeat and SOCS box-containing protein 3; ankyrin repeat-containing SOCS box protein 3; ASB3; Asb-3; EAR1; FLJ10123; FLJ10421; HREV; MGC12531; MGC132002; MGC996; THRAL
Symboles de gène ASB3
Espèces hôtes Rabbit
Immunogène Synthetic peptides corresponding to amino acids 63-75 and 166-180 of human Asb-3.
Quantité 200 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 364227, 51130, 65257
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats