missing translation for 'onlineSavingsMsg'
Learn More

Artesunate, Thermo Scientific Chemicals

Catalog No. AC460340250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460340250 25 g
AC460340050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460340250 Supplier Thermo Scientific Chemicals Supplier No. 460340250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88495-63-0
Molecular Formula C19H28O8
Molecular Weight (g/mol) 384.42
InChI Key FIHJKUPKCHIPAT-RVBFJZMGSA-N
Synonym artesunate
PubChem CID 129317578
SMILES CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC(=O)CCC(=O)O)C

Specifications

Melting Point 131.0°C to 135.0°C
Quantity 25 g
Formula Weight 384.42
Chemical Name or Material Artesunate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

RUO – Research Use Only