missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ SolventSafe Pipette Tips
GREENER_CHOICE

Catalog No. 2123618C
Change view
Click to view available options
Volume (Metric):
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
2123618C 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2123618C Supplier Thermo Scientific™ Supplier No. 5479E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform applications accurately and consistently with these Solventsafe pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ SolventSafe Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications

Description ART SolventSafe 1000E, Filtered, Sterile, Rack
Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Model ART SolventSafe 1000E
Length (English) 3.47 in.
Length (Metric) 8.83cm
Format Rack
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Product Line ART
Show More Show Less