missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Trioxide, Primary Standard, Reagents

Catalog No. 01335903
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335903 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335903 Supplier Labchem Inc Supplier No. CA2052001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Arsenic Trioxide, Primary Standard, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 101°C
Chemical Name or Material Arsenic Trioxide, Primary Standard
Color Colorless
CAS 7732-18-5, 7664-93-9, 7664-93-9
Physical Form Liquid
Quantity 1 L
Synonym Arsenic(III) Oxide
Percent Purity 1%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0685
Show More Show Less