missing translation for 'onlineSavingsMsg'
Learn More

Arsenic ICP Standard, 1000 ppm As in 3% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company PAS1KH500

Catalog No. PAS1KH500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Arsenic ICP Standard
7440-38-2 , 7647-01-0 , 7732-18-5
0.0,3.5,98.0
MFCD00085309
[As]
74.92
CHEBI:27563
Natural Poly Bottle
0.0,2.5,96.0
As
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
1mL = 1mg As (1, 000ppm As)As in 3% HCl

Chemical Identifiers

7440-38-2
74.92
RQNWIZPPADIBDY-UHFFFAOYSA-N
CHEBI:27563
[As]
As
MFCD00085309
5359596
arsenic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents

Promotions

Promotions