missing translation for 'onlineSavingsMsg'
Learn More

Arsenic AA Standard, 1000 ppm As in 3% HNO3, Ricca Chemical

Catalog No. p-3255639
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAS1KN100 100 mL Natural LDPE Bottle
AAS1KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAS1KN100 Supplier Ricca Chemical Company Supplier No. AAS1KN100
Only null left

Chemical Identifiers

CAS 1327-53-3 , 7697-37-2 , 1327-53-3 , 7732-18-5
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm As
CAS Min % 93.0, 4.62, 0.12
CAS Max % 97.0, 4.81, 0.13
Equivalence 1 mL = 1 mg As
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material Arsenic AA Standard
Show More Show Less