missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic AA Standard, 1000 ppm As in 3% H2SO4, Ricca Chemical
Spécifications
Spécifications
| Nom ou substance chimique | Arsenic AA Standard |
| Emballage | Natural Poly Bottle |
| CAS | 1327-53-3, 7664-93-9, 1327-53-3, 7732-18-5 |
| Notes sur la concentration ou la composition | 1,000ppm As |
| CAS % minimum | 93.0, 5.12, 0.12 |
| CAS % maximal | 97.0, 5.33, 0.13 |
| Équivalent | 1 mL = 1 mg As |