missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1268, Restek™

Numéro de catalogue. 06720009
Encompass
Modifier l'affichage
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
06720009 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 06720009 Fournisseur Chromatographic Code fournisseur RK 32410

Identifiants chimiques

CAS 11100-14-4

Spécifications

Packaging Ampul
Vapor Density 2.97 (Air = 1)
Flash Point -8°C
Specific Gravity 0.672 g/cm3 at 15°C
Concentration 1000 μg/mL in hexane
UN Number UN1208
Melting Point -95°C
Type Single-component Standard
Quantity 1 mL
Hazard Category Danger
Hazard Statement Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement Keep away from heat/sparks/open flames/hot surfaces. 
No smoking. Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilation and lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe dust/fume/gas/mist/vapours/spray.
Wash hands and skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
ShelfLife 6 months
EINECSNumber 203-777-6