missing translation for 'onlineSavingsMsg'
Learn More

Arginine, USP, 98.5-101.5%, Spectrum™ Chemical

A1334, 74-79-3, C6H14N4O2

$153.77 - $16,857.63

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18600754
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $153.77
Only null left
 
18600755
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $294.67
Only null left
 
18600756
View Documents Promotion Details
N/A 1 kg Amber Glass Bottle
Each for $1,136.65
Only null left
 
18600757
View Documents Promotion Details
N/A 12 kg Poly Pail
Each for $6,269.88
Only null left
 
18600758
View Documents Promotion Details
N/A 25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18600759
View Documents Promotion Details
N/A 50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Arginine, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

74-79-3
174.20
ODKSFYDXXFIFQN-UHFFFAOYNA-N
NC(CCCN=C(N)N)C(O)=O
C6H14N4O2
MFCD00002635
2-amino-5-[(diaminomethylidene)amino]pentanoic acid

Specifications

74-79-3
0.05%
0.5%
MFCD00002635
0.03%
NC(CCCN=C(N)N)C(O)=O
174.20
USP
100%
0.3%
C6H14N4O2
+26.3° to +27.7°
ODKSFYDXXFIFQN-UHFFFAOYNA-N
2-amino-5-[(diaminomethylidene)amino]pentanoic acid
98.5 to 101.5%
Videos
SDS
Documents

Documents