missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginine, USP, 98.5-101.5%, Spectrum™ Chemical
A1334, 74-79-3, C6H14N4O2
Supplier: Spectrum Chemical Mfg Cor A13341KG
| Quantity | 1 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Arginine, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 74-79-3 | |
| 174.20 | |
| ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
| NC(CCCN=C(N)N)C(O)=O |
| C6H14N4O2 | |
| MFCD00002635 | |
| 2-amino-5-[(diaminomethylidene)amino]pentanoic acid |
Specifications
| 74-79-3 | |
| 0.05% | |
| 0.5% | |
| Amber Glass Bottle | |
| +26.3° to +27.7° | |
| ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
| 2-amino-5-[(diaminomethylidene)amino]pentanoic acid | |
| 1 kg | |
| USP |
| 100% | |
| 0.3% | |
| C6H14N4O2 | |
| MFCD00002635 | |
| 0.03% | |
| NC(CCCN=C(N)N)C(O)=O | |
| 174.20 | |
| 98.5 to 101.5% |