missing translation for 'onlineSavingsMsg'
Learn More

Arginine, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18600759
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600759 50 kg Fiber Drum
18600754 25 g Amber Glass Bottle
18600755 100 g Amber Glass Bottle
18600756 1 kg Amber Glass Bottle
18600757 12 kg Poly Pail
18600758 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600759 Supplier Spectrum Chemical Mfg Cor Supplier No. A133450KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A1334, 74-79-3, C6H14N4O2

Spectrum™ Chemical Arginine, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

CAS Max % 100%
Chloride 0.05%
Ignition Residue 0.3%
Loss on Drying 0.5%
Packaging Fiber Drum
Specific Rotation +26.3° to +27.7°
Sulfate 0.03%
Quantity 50 kg
Percent Purity 98.5 to 101.5%
Grade USP