missing translation for 'onlineSavingsMsg'
Learn More

Arginine, USP, 98.5-101.5%, Spectrum™ Chemical

A1334, 74-79-3, C6H14N4O2

Supplier:  Spectrum Chemical Mfg Cor A133425KGBL

Quantity 25 kg
Packaging Fiber Drum

 View more versions of this product

Catalog No. 18600758

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Spectrum™ Chemical Arginine, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

74-79-3
174.20
ODKSFYDXXFIFQN-UHFFFAOYNA-N
NC(CCCN=C(N)N)C(O)=O
C6H14N4O2
MFCD00002635
2-amino-5-[(diaminomethylidene)amino]pentanoic acid

Specifications

74-79-3
0.05%
0.5%
Fiber Drum
+26.3° to +27.7°
ODKSFYDXXFIFQN-UHFFFAOYNA-N
2-amino-5-[(diaminomethylidene)amino]pentanoic acid
25 kg
USP
100%
0.3%
C6H14N4O2
MFCD00002635
0.03%
NC(CCCN=C(N)N)C(O)=O
174.20
98.5 to 101.5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions