missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginine Hydrochloride, USP, 98.5-101.5%, Spectrum™ Chemical
A1337, 1119-34-2, C6H14N4O2·HCl
$198.31 - $19,489.52
Chemical Identifiers
| CAS | 1119-34-2 |
|---|---|
| Molecular Formula | C6H15ClN4O2 |
| Molecular Weight (g/mol) | 210.66 |
| InChI Key | KWTQSFXGGICVPE-WCCKRBBISA-N |
| IUPAC Name | hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride |
| SMILES | [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18600749
|
N/A | 25 g | Poly Bottle |
Each for $198.31
|
|
||||
|
18600750
|
N/A | 100 g | Poly Bottle |
Each for $305.46
|
|
||||
|
18600751
|
N/A | 1 kg | Poly Bottle |
Each for $1,207.65
|
|
||||
|
18600752
|
N/A | 12 kg | Poly Pail |
Each for $6,754.25
|
|
||||
|
18600753
|
N/A | 50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Arginine Hydrochloride, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 1119-34-2 | |
| 210.66 | |
| hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride |
| C6H15ClN4O2 | |
| KWTQSFXGGICVPE-WCCKRBBISA-N | |
| [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O |
Specifications
| 1119-34-2 | |
| 0.1% | |
| C6H15ClN4O2 | |
| 0.03% | |
| [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O | |
| 210.66 | |
| USP |
| 100% | |
| 0.2% | |
| +21.4° to +23.6° | |
| KWTQSFXGGICVPE-WCCKRBBISA-N | |
| hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride | |
| 98.5 to 101.5% |