missing translation for 'onlineSavingsMsg'
Learn More

Arginine Hydrochloride, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18600749
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600749 25 g Poly Bottle
18600750 100 g Poly Bottle
18600751 1 kg Poly Bottle
18600752 12 kg Poly Pail
18600753 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600749 Supplier Spectrum Chemical Mfg Cor Supplier No. A133725GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1337, 1119-34-2, C6H14N4O2·HCl

Spectrum™ Chemical Arginine Hydrochloride, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.66
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
IUPAC Name hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
SMILES [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.2%
Packaging Poly Bottle
Specific Rotation +21.4° to +23.6°
Sulfate 0.03%
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade USP