missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Argatroban

Catalog No. AC466390010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466390010 1 g
AC466392500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466390010 Supplier Thermo Scientific Chemicals Supplier No. 466390010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74863-84-6
Molecular Formula C23H36N6O5S

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: insoluble.
Melting Point 188.0°C to 189.0°C
Formula Weight 508.64
InChI Key KXNPVXPOPUZYGB-VSVYTNTFSA-N
SMILES C[C@@H]1CCN([C@H](C1)C(O)=O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=CC2=C1NCC(C)C2
IUPAC Name (2R,4R)-1-{5-[(diaminomethylidene)amino]-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamido)pentanoyl}-4-methylpiperidine-2-carboxylic acid
Molecular Weight (g/mol) 508.64
Chemical Name or Material Argatroban

RUO – Research Use Only