accessibility menu, dialog, popup

UserName

CAS RN 74863-84-6

IUPAC Name: (2R,4R)-1-{5-[(diaminomethylidene)amino]-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamido)pentanoyl}-4-methylpiperidine-2-carboxylic acid
Synonyms: argatroban
Molecular Weight (g/mol): 508.64
Molecular Formula: C23H36N6O5S
InChi Key: KXNPVXPOPUZYGB-VSVYTNTFSA-N
SMILES: C[C@@H]1CCN([C@H](C1)C(O)=O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=CC2=C1NCC(C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)