missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Arctigenin

Catalog No. p-7214720
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466040010 1 g
AC466042500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466040010 Supplier Thermo Scientific Chemicals Supplier No. 466040010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7770-78-7
Molecular Formula C21H24O6

Specifications

Quantity 1 g
Melting Point 100.0°C
Formula Weight 372.42
InChI Key NQWVSMVXKMHKTF-JKSUJKDBSA-N
SMILES COC1=CC=C(C[C@H]2COC(=O)[C@@H]2CC2=CC=C(O)C(OC)=C2)C=C1OC
IUPAC Name (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]oxolan-2-one
Molecular Weight (g/mol) 372.42
Chemical Name or Material (-)-Arctigenin

RUO – Research Use Only