missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Arctigenin

Catalog No. ac466040010
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AC466040010 1 g
AC466042500 250 mg
2 options

Catalog No. AC466040010

Supplier: Thermo Scientific Chemicals 466040010

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7770-78-7
Molecular Formula C21H24O6

Specifications

Quantity 1 g
Melting Point 100.0°C
Formula Weight 372.42
InChI Key NQWVSMVXKMHKTF-JKSUJKDBSA-N
SMILES COC1=CC=C(C[C@H]2COC(=O)[C@@H]2CC2=CC=C(O)C(OC)=C2)C=C1OC
IUPAC Name (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]oxolan-2-one
Molecular Weight (g/mol) 372.42
Chemical Name or Material (-)-Arctigenin

RUO – Research Use Only