missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Arctigenin

Catalog No. AC466042500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466042500 250 mg
AC466040010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466042500 Supplier Thermo Scientific Chemicals Supplier No. 466042500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7770-78-7
Molecular Formula C21H24O6

Specifications

Quantity 250 mg
Melting Point 100.0°C
Formula Weight 372.42
Chemical Name or Material (-)-Arctigenin

RUO – Research Use Only