missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Arctic Express™ Dual Storage Systems

Catalog No. 11675903
Change view
Click to view available options
Capacity (Metric):
10 L
18.5 L
28 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11675903 28 L
11675900 10 L
11675901 18.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11675903 Supplier Thermo Scientific™ Supplier No. CK50922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.

Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.

Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a 'can and cane' storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.

Specifications

Type Manual Fill LN2 Dewar
Capacity (Metric) 28 L
No. of Doors 1
Model Arctic Express Dual 28
Static Holding Time 21 shipper/50 lab
Diameter (English) 18.2 in.
Diameter (English) Inside Neck 3.8 in.
Diameter (Metric) 46.2 cm
Diameter (Metric) Inside Neck 9.7 cm
Vial Capacity 630 x 2mL