missing translation for 'onlineSavingsMsg'
Learn More

α-Arbutin, ≥99% (HPLC and GC), MilliporeSigma™ Supelco™

Catalog No. 111015667
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015667 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015667 Supplier Merck Emd Millipore Supplier No. 03338100MG
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 84380-01-8
Linear Formula C12H16O7
MDL Number MFCD09838262
Synonym 4-Hydroxyphenyl alpha-D-glucopyranoside; Hydroquinone O-alpha-D-glucopyranoside
Type Analytical Standard
Quantity 100 mg
Formula Weight 272.25
Percent Purity ≥99% (HPLC and GC)
Grade Analytical Standard