missing translation for 'onlineSavingsMsg'
Learn More

Arachidonic Acid 98.0+%, TCI America™

Catalog No. A0781500MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0781500MG 500 mg
A0781100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0781500MG Supplier TCI America Supplier No. A0781500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 506-32-1
Molecular Formula C20H32O2
Molecular Weight (g/mol) 304.474
MDL Number MFCD00004417
InChI Key YZXBAPSDXZZRGB-DOFZRALJSA-N
Synonym arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid
PubChem CID 444899
ChEBI CHEBI:15843
IUPAC Name (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid
SMILES CCCCCC=CCC=CCC=CCC=CCCCC(=O)O

Specifications

Color Yellow
Quantity 500 mg
Formula Weight 304.47
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Arachidonic Acid
EINECSNumber (2)-0609
RTECSNumber CE6675000
TSCA No
Recommended Storage Freezer