missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AR-A 014418

Catalog No. 396610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
396610 10 mg
396650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 396610 Supplier Tocris Bioscience™ Supplier No. 3966/10*

Selective GSK-3 inhibitor

Selective glycogen synthase kinase 3 (GSK-3) inhibitor (IC50 = 104 nM). Exhibits specificity for GSK-3 over cdk2 and cdk5 (IC50 values are > 100 μM) and over 26 other kinases. Inhibits β-amyloid-mediated neurodegeneration in hippocampal slices.

Chemical Identifiers

CAS 487021-52-3
Molecular Formula C12H12N4O4S

Specifications

Quantity 10 mg
Formula Weight 308.31
Purity >99%
Molecular Weight (g/mol) 308.312
Chemical Name or Material AR-A 014418
Recommended Storage Store at +4°C