CAS RN 487021-52-3
IUPAC Name:
1-[(4-methoxyphenyl)methyl]-3-(5-nitro-1,3-thiazol-2-yl)urea
Synonyms:
1-[(4-methoxyphenyl)methyl]-3-(5-nitro-1,3-thiazol-2-yl)urea
Molecular Weight (g/mol):
308.31
Molecular Formula:
C12H12N4O4S
InChi Key:
YAEMHJKFIIIULI-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CNC(=O)NC2=NC=C(S2)[N+]([O-])=O)C=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals GSK-3beta Inhibitor VIII
CAS: 487021-52-3 Molecular Formula: C12H12N4O4S Synonym: AR-A014418
2
R&D Systems™ AR-A 014418
CAS: 487021-52-3 Molecular Formula: C12H12N4O4S