missing translation for 'onlineSavingsMsg'
Learn More

Hygiena Aquasnap™ ATP Water Testing Device

Catalog No. 14037769
Change view
Click to view available options
For Use With (Application):
Detects Free ATP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
14037769 Detects Free ATP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14037769 Supplier Hygiena Supplier No. AQ100FX
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

Luminometer

User-friendly, self-contained ATP water sampling device used with the Hygiena luminometers

AquaSnap is easy to use, economical, and gives real time results. The specifically designed dipper tip collects 100μl of water ensuring consistent sample collection.

AquaSnap Total measures both free ATP in solution and microbial ATP. AquaSnap Total contains a detergent to release ATP that is bound to microbial or organic matter and inside microbes. AquaSnap Free measures dissolved ATP that is free in solution. It is intended for use in food and beverage processing to detect organic contamination in rinse water samples from CIP systems, tanks and pipes. The difference between Total and Free ATP provides a measure of microbial contamination in the sample.

  • Patented Snap-Valve™ technology and self-contained liquid stable reagent minimizes costs and maximizes performance
  • Write-on swab label
  • Results in 15 seconds
  • Unprecedented accuracy and reproducibility
  • Honeycomb-shaped collection tip collects 100μL of sample - no pipetting
  • Compatible with SystemSURE Plus and EnSURE luminometers

Specifications

For Use With (Application) Detects Free ATP
Required Accessories Luminometer
Storage Requirements 2°C to 8°C