missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Electrolyte CG, for Karl Fischer Titration By Coulometry, Fisher Chemical

Catalog No. AL256025
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL256025 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AL256025 Supplier Thermo Fisher Scientific Supplier No. AL256025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

25mL, Vial

  • Catholyte solution for use in conventional cells with a diaphragm
  • Reagent required to complete the electrochemical reaction in the cathode compartment for coulometric analysis
  • Contains methanol as a solvent

Specifications

Chemical Name or Material Aqualine™ Electrolyte CG
Name Note Karl Fischer Reagent
Boiling Point 64°C
Color Colorless
CAS 7446-09-5, 108-75-8, 7553-56-2
Physical Form Liquid
Quantity 25 mL
Packaging Vial
MDL Number 3097388