missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Complete 1, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Catalog No. K19001
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
K19001 1 L Amber glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K19001 Supplier Fisher Chemical Supplier No. K190015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 1.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free

Specifications

Chemical Name or Material Aqualine™ Complete 1
Name Note Karl Fischer Reagent
Color Colorless
CAS 7446-09-5, 288-32-4, 111-90-0, 7553-56-2
Physical Form Liquid
Quantity 1 L
Molecular Formula Solution
Packaging Amber glass bottle
MDL Number 81734