Learn More
Microsolv Technology Aqr Snapcap 11Mm Preslit 100Pk DFS Item
Supplier:  Microsolv Technology Corporation 9509C30CB
Caps, Snap Top, with Silicone Rubber / PTFE, ultra-pure, Pre-Slit Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm, Snap Top autosampler vials. AQR Brand. 100/PK.
Catalog No. NC1310289
Only null left
Explore available promotions