missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apronal 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 196°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 184.24 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Apronal |
Safety and Handling
| EINECSNumber | (9)-0107 |
| RTECSNumber | YT5775000 |
| TSCA | No |